J.T.Baker® DUAL-TINT®, pH Indicator Papers
Catalog # 2873-01
Supplier: Avantor
Specifications
- pH gradation:1
- pH range:1 - 12
- 説明:DUAL-TINT® Refill
- Seq:02
- Pk:1
Specifications
About this item
DUAL-TINT® pH papers offer easy-to-read, precise pH indication in a rolled 17-foot strip wide in a refillable dispenser with colour scale.
Two indicator zones are on one strip, separated by a white center division to give you clear colour gradations and reliable pH values.
Ordering information: Refills are avaialble for the dispenser in packs of three rolls.