Avantor® ACE® Generix®, Analytical HPLC Columns, 5 µm
Supplier: Avantor
Certificates
About this item
These high performance HPLC columns contain high purity spherical silica.
- Ultra-narrow particle size distribution for excellent column efficiency
- Sharp, highly symmetrical peaks for acidic, basic and neutral analytes
- Excellent column to column and batch to batch reproducibility
Each column is individually tested and supplied with a test chromatogram.
Specifications
- Catalog No:
- GEN-5PHEX-2546
- GEN-5SIL-7503
- GEN-5SIL-0503
- GEN-5SIL-2502
- GEN-5SIL-1502
- GEN-5SIL-1202
- GEN-5SIL-1002
- GEN-5SIL-7502
- GEN-5SIL-2546
- GEN-5SIL-0502
- GEN-5SIL-1546
- GEN5PHEX2546/3
- GEN-5SIL-1246
- GEN5PHEX1546/3
- GEN-5SIL-1046
- GEN5PHEX0502/3
- GEN-5SIL-7546
- GEN-5SIL-0546
- GEN-5SIL-2504
- GEN-5SIL-1504
- GEN-5SIL-1204
- GEN-5SIL-1004
- GEN-5SIL-7504
- GEN-5SIL-0504
- GEN5SIL2546/3
- GEN-5SIL-2503
- GEN-5PHEX-1546
- GEN5SIL1546/3
- GEN-5SIL-1503
- GEN5SIL0502/3
- GEN-5SIL-1203
- GEN-5SIL-1003
- Description:
- Phenyl-Hexyl
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- Phenyl-Hexyl
- SIL
- Phenyl-Hexyl
- SIL
- Phenyl-Hexyl
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- Phenyl-Hexyl
- SIL
- SIL
- SIL
- SIL
- SIL
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 15.2%
- –
- –
- –
- –
- –
- –
- –
- –
- –
- –
- 15.2%
- –
- 15.2%
- –
- 15.2%
- –
- –
- –
- –
- –
- –
- –
- –
- –
- –
- 15.2%
- –
- –
- –
- –
- –
- Endcapped:
- Yes
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Yes
- No
- Yes
- No
- Yes
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Yes
- No
- No
- No
- No
- No
- pH range:
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- Surface area:
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- USP code:
- L11
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L11
- L3
- L11
- L3
- L11
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L11
- L3
- L3
- L3
- L3
- L3
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Specifications
Guard cartridges - guard holder required