Orders LinkContactUsLinkComponent
VWR®, エコノミースピルキット
VWR®, エコノミースピルキット
VWR®, エコノミースピルキット
VWR®, エコノミースピルキット
VWR®, エコノミースピルキット
These economy spill kits combine absorbents and instructions in a space-saving container. They are ideal for workers on the go like truck drivers, field workers, etc.
  • Improve worker productivity and efficiency
  • Fast spill response
  • Yellow PVC bag offers high visibility spill control storage
  • Highly versatile, surfactant treated, fire resistance
  • Water resistant, lightweight bag keeps sorbents dry
Brightly coloured absorbent, picks up a wide range of aggressive chemicals while remaining chemically inert. White absorbents pick up petroleum-based fluids, paints and non water soluble chemicals while repelling water - great for indoor and outdoor use. Grey absorbents pick up water-based, petroleum-based and non aggressive, chemical fluids.
Ordering information: Every kit is supplied with 25 pads (410×510 mm), 4 socks (Ø 76×1220 mm) and 1 instruction sheet.
Packaging: Packed in a durable, see-through PVC bag with shoulder strap.
Product Family Options
  • 説明
    Absorbency
    W×D×H
  • Economy spill kit, chemical
    32 l
    406,4×508,0×101,6 mm
    1 * 1 items
    1 * 1 items
  • Economy spill kit, maintanence
    32 l
    406,4×508,0×101,6 mm
    1 * 1 items
    1 * 1 items
  • Economy spill kit, oil
    32 l
    406,4×508,0×101,6 mm