Orders LinkContactUsLinkComponent
VWR® VistaVision, Microscope slides, colour-coded
VWR® VistaVision, Microscope slides, colour-coded
Supplier:   VWR Collection
VWR® VistaVision, Microscope slides, colour-coded
Supplier:  VWR Collection
Certificates
VWR® VistaVision, Microscope slides, colour-coded
3 Options Available Below
Product Details & Documents
Bright printed colours of these microscope slides provide an ideal marking area and enable easy identification. The ultra-thin colour frosted end facilitates processing on automated systems.
  • Manufactured using high quality, white soda lime glass
  • Pre-cleaned and ready for use
  • Autoclavable
  • Colour coded ends facilitate sorting and identification
  • Uniform size and thickness
Certifications: ISO 8037/1
Product Family Options
Product Information
  • 説明
    Colour
    厚さ
    長さ×幅
    Availability
    Price
  • Microscopy slides, premium, square corners
    White
    1,0 mm
    75×25 mm
    Log in for availability
    Log in for availability
    1 * 72 items
    1 * 72 items - $46.76
    $46.76
    1 * 72 items
       $46.76
  • Microscopy slides, premium, square corners
    Blue
    1,0 mm
    75×25 mm
    Log in for availability
    Log in for availability
    1 * 1.440 items
    1 * 1.440 items
  • Microscopy slides, premium, square corners
    Yellow
    1,0 mm
    75×25 mm
    Log in for availability
    Log in for availability
    1 * 72 items
    1 * 72 items - $46.76
    $46.76
    1 * 72 items
       $46.76