Orders LinkContactUsLinkComponent
シューズカバー、SMS
シューズカバー、SMS
Supplier:   VWR Collection
シューズカバー、SMS
Supplier:  VWR Collection
Certificates
シューズカバー、SMS
シューズカバー、SMS
3 Options Available Below
Product Details & Documents
These disposable sterile boot covers are manufactured from 60 g/m² spunbond meltblown spunbond SMS fabric and feature a 250 g/m² PVC sole.
  • Bonded seams
  • Elastic bands for a secure fit
  • Slip resistant sole
  • Sterility Assurance Level (SAL) of 10⁻⁶
Certifications: ISO 9001, ISO13485, ISO14001, GMP
Packaging: Boot covers are vacuum packed in pairs.
Product Family Options
Product Information
  • 説明
    Colour
    高さ
    Availability
    Price
  • Shoe covers, medium
    White
    726 mm
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items
  • Shoe covers, universal
    White
    901 mm
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items
  • Shoe covers, x-large
    White
    965 mm
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items