保護ガウン、SPP
About this item
Economical gowns are manufactured from a strong, lightweight, breathable and non-linting spunbonded polypropylene (SPP) fabric.
- Provide first line barrier
- Elastic cuffs and ties at the neck and waist ensure secure closure with
- Serged seams
- No pockets
Gowns protect against dirt, grime and certain dry particulates in nonhazardous environments.
: Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
2 Options Available Below
Product Details & Documents
Economical gowns are manufactured from a strong, lightweight, breathable and non-linting spunbonded polypropylene (SPP) fabric.
- Provide first line barrier
- Elastic cuffs and ties at the neck and waist ensure secure closure with
- Serged seams
- No pockets
Gowns protect against dirt, grime and certain dry particulates in nonhazardous environments.
: Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
Specifications for Product Family
Specifications
- Catalog No:
- VWRU414004-467
- VWRU414004-468
- Fabric:
- SPP
- SPP
- Pockets:
- Without pockets
- Without pockets
- Seam type:
- Serged
- Serged
- Garment application:
- Medical|Clinical
- Medical|Clinical
Specifications
Product Family Options
Product Information
- Colour衣服のサイズAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.VWRU414004-467FabricSPPPocketsWithout pocketsSeam typeSergedGarment applicationMedical|Clinical - Specifications:
More
Specifications
Cat. No.VWRU414004-468FabricSPPPocketsWithout pocketsSeam typeSergedGarment applicationMedical|Clinical