VWR®, Measuring Cylinders, Borosilicate Glass, Class B, Calibrated to Deliver
Supplier: VWR Collection
Certificates
About this item
Borosilicate glass. Cylinder sizes from 25 ml to 2000 ml feature a hexagonal base and yellow bumper.
- Borosilicate glass fabrication
- Calibrated at 20 °C to deliver
- Durable white colour scale printing for long life in laboratory environment
'TD' appears on each cylinder and indicates the cylinder is calibrated to deliver.
Certifications: Made from ASTM E438 Type 1 Class A borosilicate 3,3 glass. As per ASTM Specification E1272 Class B requirements.
Frequently Bought Together