トング、PTFEコーティング、VWR®製
229 mm
Catalog # VWRU82027-376
Supplier: VWR Collection
Jump to:
Product Details & Documents
Designed to handle hot, cold, corrosive, or sticky materials without scratching or damaging container exteriors.
- Stainless steel construction
- PTFE coating
- Nonsticking and non-corrosive
Contoured jaw enables the handling of crucibles and other objects. Tongs are used for handling chemically active materials.
Specifications
- 長さ:229 mm