Orders LinkContactUsLinkComponent
ビーカー、ローフォーム、Griffin
ビーカー、ローフォーム、Griffin
ビーカー、ローフォーム、Griffin
ビーカー、ローフォーム、Griffin
VWR® low form Griffin beakers are made from high quality Type 1, Class A, 33 expansion borosilicate glass. They feature a thick, slightly flared, beaded top with a spout designed to have excellent pouring characteristics.
  • Double capacity scale
  • Uniform wall thickness
  • Very good chemical resistance
  • High temperature resistance
Printed with white graduated scale and a durable marking area.
 : Meets ASTM E960, E960-93.
Product Family Options
  • 容量
    高さ
    外径
    目盛り範囲
  • 10 ml
    34 mm
    25 mm
    1 * 12 items
    1 * 12 items - $58.11
    $58.11
    1 * 12 items
       $58.11
  • 50 ml
    53 mm
    42 mm
    10 - 40 ml
    1 * 12 items
    1 * 12 items - $45.67
    $45.67
    1 * 12 items
       $45.67
  • 100 ml
    70 mm
    50 mm
    20 - 80 ml
    1 * 12 items
    1 * 12 items - $50.57
    $50.57
    1 * 12 items
       $50.57
  • 150 ml
    81 mm
    57,5 mm
    20 - 140 ml
    1 * 12 items
    1 * 12 items - $58.34
    $58.34
    1 * 12 items
       $58.34
  • 250 ml
    88 mm
    68 mm
    25 - 200 ml
    1 * 12 items
    1 * 12 items - $63.56
    $63.56
    1 * 12 items
       $63.56
  • 400 ml
    106 mm
    77 mm
    25 - 325 ml
    1 * 12 items
    1 * 12 items - $65.08
    $65.08
    1 * 12 items
       $65.08
  • 600 ml
    122 mm
    88 mm
    50 - 500 ml
    1 * 6 items
    1 * 6 items - $46.60
    $46.60
    1 * 6 items
       $46.60
  • 1000 ml
    145 mm
    107 mm
    100 - 1000 ml
    1 * 6 items
    1 * 6 items - $65.08
    $65.08
    1 * 6 items
       $65.08
  • 1500 ml
    165 mm
    120 mm
    200 - 1400 ml
    1 * 4 items
    1 * 4 items - $56.61
    $56.61
    1 * 4 items
       $56.61
  • 2000 ml
    188 mm
    131 mm
    200 - 2000 ml
    1 * 4 items
    1 * 4 items - $67.02
    $67.02
    1 * 4 items
       $67.02
  • 4000 ml
    250 mm
    160 mm
    500 - 3500 ml
    1 * 1 items
    1 * 1 items - $75.38
    $75.38
    1 * 1 items
       $75.38