Orders LinkContactUsLinkComponent
メスピペットピペット、クラスA
Supplier:   VWR Collection
メスピペットピペット、クラスA
Supplier:  VWR Collection
Certificates
メスピペットピペット、クラスA
メスピペットピペット、クラスA
11 Options Available Below
Product Details & Documents
Borosilicate 3.3 expansion glass, class A.
  • Reusable
  • Colour coded for easy sorting
Calibrated at 20 °C to deliver. These pipettes are unserialised or batch certified.
 : Made from ASTM E438 Type 1 Class A borosilicate 33 expansion glass and as per ASTM specification E969. Pipettes are colour coded as per ASTM specification E1273.
Product Family Options
Product Information
  • 容量
    Tolerance
    Colour code
    長さ
    Availability
    Price
  • 1 ml
    ±0,006 ml
    Blue
    315 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items - $103.08
    $103.08
    1 * 6 items
       $103.08
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU10546-006
  • 2 ml
    ±0,006 ml
    Orange
    345 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items - $107.99
    $107.99
    1 * 6 items
       $107.99
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU10546-008
  • 3 ml
    ±0,01 ml
    Black
    360 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items - $197.78
    $197.78
    1 * 6 items
       $197.78
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU10546-010
  • 4 ml
    ±0,01 ml
    Red
    395 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items - $101.18
    $101.18
    1 * 6 items
       $101.18
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU10546-012
  • 5 ml
    ±0,01 ml
    White
    400 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items - $104.68
    $104.68
    1 * 6 items
       $104.68
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU10546-014
  • 7 ml
    ±0,01 ml
    Green
    430 mm
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU10546-018
  • 10 ml
    ±0,02 ml
    Red
    440 mm
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU10546-024
  • 15 ml
    ±0,03 ml
    Green
    470 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items - $228.55
    $228.55
    1 * 6 items
       $228.55
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU10546-026
  • 20 ml
    ±0,03 ml
    Yellow
    510 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items - $230.54
    $230.54
    1 * 6 items
       $230.54
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU10546-028
  • 30 ml
    ±0,03 ml
    Black
    530 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items - $236.99
    $236.99
    1 * 6 items
       $236.99
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU10546-032
  • 50 ml
    ±0,05 ml
    Red
    550 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items - $252.16
    $252.16
    1 * 6 items
       $252.16
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU10546-036