Orders LinkContactUsLinkComponent
クリーンルーム用ワイパー, ポリエステル
クリーンルーム用ワイパー, ポリエステル
クリーンルーム用ワイパー, ポリエステル
クリーンルーム用ワイパー, ポリエステル
Durable wipers resist fibre release and abrasion under rigorous use, even when wiping rough surfaces.
  • 100% virgin, interlock knit polyester
  • Continuous filament
  • No-run material
  • Laser cut
  • Sealed edges
Laser cut wipers with sealed edges limit particles and fibres to an absolute minimum, ensuring excellent performance in environments that demand low levels of contaminants.
Made from 100% virgin-grade, continuous-filament polyester in a double-knit, no-run interlock pattern to provide maximum cleanliness and absorbency. Designed for cleaning surfaces and controlling spills in ISO Class 3 (FED-STD-209E Class 1/M1.5) or less critical environments.
 : Laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom. Wipers comply with RoHS 2002/95/EC.
 : Packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom. Wipers are vacuum-packed in solvent-safe, cleanroom-compatible polyethylene with a label on each inner bag.
Product Family Options
  • Colour
    長さ×幅
    Packed
  • White
    229×229 mm
    150/bag
    1 * 1.200 items
    1 * 1.200 items
  • White
    305×305 mm
    150/bag
    1 * 900 items
    1 * 900 items