VWR® CryoPro® , Plastic storage boxes
Large cryogenic freezer storage box, 51 mm, 133L×133D×510H mm
Catalog # VWRU89214-760
Supplier: VWR Collection
Jump to:
Product Details & Documents
Polycarbonate construction for use in mechanical and LN₂ freezers.
- Grid format for lids
- Boxes feature drain slots for safe liquid nitrogen storage
Lids come printed with 81 and 100 cell grids to assist with inventory control.
Specifications
- W×D×H ext.:133L×133D×510H mm
- 説明:Large cryogenic freezer storage box
- 高さ:51 mm
- Seq:02