作業着、ジッパー付き 最大
VWR® Maximum SF frocks are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness and comfort properties.
- Snap collar, raglan sleeves, zip closure, elasticated cuffs
- Superior fluid and particulate barrier
- Lowest level of particle shedding: Level 1 Helmke drum classification
- Light chemical splash resistance
- Improved breathability to ensure user comfort
- Soft, cloth-like fabric with maximum tear strength
The frocks provide barrier protection against fluids, biologicals and particulates. A high vapour transmission rate keeps the wearer comfortable, even throughout extended use. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental clean-up applications.
: Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
VWR® Maximum SF frocks are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness and comfort properties.
- Snap collar, raglan sleeves, zip closure, elasticated cuffs
- Superior fluid and particulate barrier
- Lowest level of particle shedding: Level 1 Helmke drum classification
- Light chemical splash resistance
- Improved breathability to ensure user comfort
- Soft, cloth-like fabric with maximum tear strength
The frocks provide barrier protection against fluids, biologicals and particulates. A high vapour transmission rate keeps the wearer comfortable, even throughout extended use. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental clean-up applications.
: Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
- Catalog No:
- VWRU414004-696
- VWRU414004-697
- VWRU414004-698
- VWRU414004-699
- VWRU414004-700
- VWRU414004-701
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Pockets:
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Garment application:
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
Product Family Options
- Colour衣服のサイズ
Specifications
Cat. No.VWRU414004-696Closure typeZip closurePocketsWithout pocketsGenderUnisexGarment applicationCleanroom|LaboratorySpecifications
Cat. No.VWRU414004-697Closure typeZip closurePocketsWithout pocketsGenderUnisexGarment applicationCleanroom|LaboratorySpecifications
Cat. No.VWRU414004-698Closure typeZip closurePocketsWithout pocketsGenderUnisexGarment applicationCleanroom|LaboratorySpecifications
Cat. No.VWRU414004-699Closure typeZip closurePocketsWithout pocketsGenderUnisexGarment applicationCleanroom|LaboratorySpecifications
Cat. No.VWRU414004-700Closure typeZip closurePocketsWithout pocketsGenderUnisexGarment applicationCleanroom|LaboratorySpecifications
Cat. No.VWRU414004-701Closure typeZip closurePocketsWithout pocketsGenderUnisexGarment applicationCleanroom|Laboratory