Orders LinkContactUsLinkComponent
粉体漏斗、ガラス、VWR®製
粉体漏斗、ガラス、VWR®製
粉体漏斗、ガラス、VWR®製
粉体漏斗、ガラス、VWR®製
Made from Type 1 Class A borosilicate 3.3 expansion glass.
  • Constructed with a heavy, uniform wall
  • Fire-polished rim and stem to provide long service life
  • Chemical resistant
  • White printing
Funnels comes with short, wide stem, and are useful for adding powdered chemical to reaction vessels.
 : In acc. with ASTM E-1095 standards.
Product Family Options
  • Funnel O-Ø
    Stem O-Ø
  • 60 mm
    20 mm
    1 * 6 items
    1 * 6 items - $172.15
    $172.15
    1 * 6 items
       $172.15
  • 80 mm
    25 mm
    1 * 6 items
    1 * 6 items - $157.04
    $157.04
    1 * 6 items
       $157.04
  • 100 mm
    25 mm
    1 * 6 items
    1 * 6 items - $196.07
    $196.07
    1 * 6 items
       $196.07
  • 150 mm
    31 mm
    1 * 2 items
    1 * 2 items - $345.71
    $345.71
    1 * 2 items
       $345.71