Orders LinkContactUsLinkComponent
ブーツ付きオーバーオール, SMS
ブーツ付きオーバーオール, SMS
Supplier:   VWR Collection
ブーツ付きオーバーオール, SMS
Supplier:  VWR Collection
Certificates
4 Options Available Below
Product Details & Documents
These overalls with integrated shoe covers are manufactured from a unique multilayered spunbonded-meltblown-spunbonded (SMS) fabric that offers high tensile strength while remaining soft, comfortable and breathable. The material is low-linting and resistant to tears and punctures.
  • Light fluid and particulate barrier
  • Excellent breathability
  • Inset sleeves, front zipper, tapered collar
  • Elastic cuffs, ankles, and back
  • No pockets
  • Serged seams
These static-dissipative garments act as a fluid and particulate barrier and provide light chemical splash resistance. Ideal for use in high-class protection areas of medical, industrial, laboratory, electronic, and pharmaceutical environments.
Certifications: Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
Specifications for Product Family
Specifications
  • Catalog No:
  • VWRU414004-688
  • VWRU414004-689
  • VWRU414004-690
  • VWRU414004-691
  • Garment application:
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Bacterial filtration efficiency:
  • 0,9
  • 0,9
  • 0,9
  • 0,9
Specifications
Product Family Options
Product Information
  • Colour
    衣服のサイズ
    Availability
    Price
  • White
    M
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU414004-688
    Garment application
    Laboratory
    Bacterial filtration efficiency
    0,9
  • White
    L
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU414004-689
    Garment application
    Laboratory
    Bacterial filtration efficiency
    0,9
  • White
    XL
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU414004-690
    Garment application
    Laboratory
    Bacterial filtration efficiency
    0,9
  • White
    XXL
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU414004-691
    Garment application
    Laboratory
    Bacterial filtration efficiency
    0,9