VWR® smartPans™, 秤量皿
Supplier: VWR Collection
Certificates
About this item
Cahn's style weighing pans compatible with most available analysers and microbalances.
- Made of Ultra High Vacuum (UHV) foil
- Wettable and completely free from oils (normally found on other grades of aluminum foil)
Suitable for organic, micro elementary and thermal analysis. Also convenient for pharmaceutical applications.
Certifications: ASTM B-479 3.1.4 and 10.3.1.
Packaging: In easy-to-use polypropylene flip-top vials.
Specifications
- Catalog No:
- 611-3834
- 611-3835
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing