Hematoxylin, BAKER ANALYZED®, Certified Stain for histology, for cytology, J.T.Baker®
M906-03
: Avantor
:
- パックタイプ:Glass bottle, amber, wide mouth, black T/E cap
- Pk:25 g
- Seq:3308
- Biological Test:Passes Test
:
:302.28 g/mol
:13,04654
:291…293 °C
:Ambient
:208-237-3
:517-28-2
:C₁₆H₁₄O₆
:MFCD00078111