J.T.Baker® PharmaTest™, pH Buffer, Ammonia-Ammonium Chloride
5904-02
: Avantor
:
- パックタイプ:PE bottle, narrow mouth
- pH buffer type:Ammonia
- Pk:1 L
- 説明:Ammonia-ammonium chloride buffer
- pH value:10,08 - 10,12
: Meets test solution (TS) specifications for testing USP/NF monographs.