Orders LinkContactUsLinkComponent
J.T.Baker® BAKER ANALYZED®, Buffer Solutions
J.T.Baker® BAKER ANALYZED®, Buffer Solutions
 :   Avantor
J.T.Baker® BAKER ANALYZED®, Buffer Solutions
 :  Avantor
J.T.Baker® BAKER ANALYZED®, Buffer Solutions
J.T.Baker® BAKER ANALYZED®, Buffer Solutions
Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilise quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardisation of solutions, helping streamline high-volume analytical work in applications such as quality control.
Contain 0,01% Sodium benzoate.
Certifications: This buffer is traceable to NIST Standard reference buffer.
Product Family Options
  • pH value
    pH accuracy
    pH buffer type
    Reference temperature
    パックタイプ
    Pk
  • 4,00
    ±0,02
    Phthalate
    25 °C
    Plastic bottle
    500 mL
    1 * 500 mL
    1 * 500 mL - $80.22
    $80.22
    1 * 500 mL
       $80.22
  • 4,00
    ±0,02
    Phthalate
    25 °C
    Plastic bottle
    500 mL
    1 * 500 mL
    1 * 500 mL
  • 4,00
    ±0,02
    Phthalate
    25 °C
    Plastic bottle
    1 L
    1 * 1 L
    1 * 1 L
  • 4,00
    ±0,02
    Phthalate
    25 °C
    Plastic container
    10 L
    1 * 10 L
    1 * 10 L
  • 7,00
    ±0,02
    Phosphate
    20 °C
    Plastic bottle
    500 mL
    1 * 500 mL
    1 * 500 mL - $81.52
    $81.52
    1 * 500 mL
       $81.52
  • 7,00
    ±0,02
    Phosphate
    20 °C
    Plastic bottle
    500 mL
    1 * 500 mL
    1 * 500 mL
  • 7,00
    ±0,02
    Phosphate
    20 °C
    Plastic bottle
    1 L
    1 * 1 L
    1 * 1 L
  • 7,00
    ±0,02
    Phosphate
    20 °C
    Plastic container
    10 L
    1 * 10 L
    1 * 10 L
  • 10,00
    ±0,05
    Borate
    25 °C
    Plastic bottle
    500 mL
    1 * 500 mL
    1 * 500 mL
  • 10,00
    ±0,05
    Borate
    25 °C
    Plastic bottle
    1 L
    1 * 1 L
    1 * 1 L
  • 10,00
    ±0,05
    Borate
    25 °C
    Plastic container
    10 L
    1 * 10 L
    1 * 10 L