Digital Solutions
Applications
Support
About
Japan
Orders Link
ContactUsLinkComponent
キュベット
Spectrophotometer Cuvettes
VWR®, 角型キュベット
VWR®, 角型キュベット
:
VWR Collection
VWR®, 角型キュベット
:
VWR Collection
VWR®, 角型キュベット
CoC VWR® V-1200 UV-1600PC UV-3100PC
Manual P6 Spectrophotometer
VWR-UV-3100PC-Manual-V1_EN_060910.pdf
VWR® UV-1600PC
VWR® UV-6300PC_260515
VWR® V-1200
TDS VWR Glass Cuvette,10 mm.pdf
TDS VWR Glass Cuvette,100 mm.pdf
TDS VWR Glass Cuvette,20 mm.pdf
TDS VWR Glass Cuvette,30 mm.pdf
TDS VWR Glass Cuvette,50 mm.pdf
VWR Products for Nucleic Acid Preparation
Product Family Options
説明
634-6013
Cuvette, square cuvettes, glass, path length 10 mm, 4 pcs
1 * 4 items
-
+
1 * 4 items
-
+
634-6015
Cuvette, square cuvettes, glass, path length 30 mm, 2 pcs
1 * 2 items
-
+
1 * 2 items
-
+
634-6016
Cuvette, square cuvettes, glass, path length 50 mm, 2 pcs
1 * 2 items
-
+
1 * 2 items
-
+
634-6017
Cuvette, square cuvettes, glass, path length 100 mm, 1 pcs
1 * 1 items
-
+
1 * 1 items
-
+
634-6018
Cuvette, square cuvettes, quartz, path length 10 mm, 2 pcs
1 * 2 items
-
+
1 * 2 items
-
+
634-6019
Cuvette, square cuvettes, quartz, path length 20 mm, 2 pcs
1 * 2 items
-
+
1 * 2 items
-
+
634-6020
Cuvette, square cuvettes, quartz, path length 30 mm, 2 pcs
1 * 2 items
-
+
1 * 2 items
-
+
634-6021
Cuvette, square cuvettes, quartz, path length 50 mm, 2 pcs
1 * 2 items
-
+
1 * 2 items
-
+
634-6022
Cuvette, square cuvettes, quartz, path length 100 mm, 1 pcs
1 * 1 items
-
+
1 * 1 items
-
+
VWR® P4, UV/Vis分光光度計、
#: 634-1229
: VWR Collection