VWR®, Sealing Mats for Square Well Plates
Catalog # VWRU76196-420
Supplier: VWR Collection
Specifications
- 説明:Sealing mat for 96 square well plate, sterile
- Pk:50
Specifications
About this item
Sealing mats made of EVA (Ethylene Vinyl Acetate), suitable for square well plates.
- DNAse- and RNAse-free