Orders LinkContactUsLinkComponent
VWR®, バケツライナー、滅菌
VWR®, バケツライナー、滅菌
VWR®, バケツライナー、滅菌
Supplier:  VWR Collection
Certificates
VWR®, バケツライナー、滅菌
VWR®, バケツライナー、滅菌
2 Options Available Below
Product Details & Documents
Bucket liners create an impermeable barrier between the disinfectant and receptacle.
  • LDPE/nylon blend
  • Irradiated
  • Fits B-7 series buckets
Helps ensure the integrity of a disinfectant blend.
Packaging: Individually bagged, double-bagged in groups of 6. 4 packs of 6 per case.
Product Family Options
Product Information
  • 説明
    Material
    長さ×幅
    Availability
    Price
  • Bucket liners for B-7 buckets, sterile
    LDPE/nylon
    790×480 mm
    Log in for availability
    Log in for availability
    1 * 24 items
    1 * 24 items
  • Bucket liners for B-7 buckets, sterile
    LDPE/nylon
    790×480 mm
    Log in for availability
    Log in for availability
    1 * 72 items
    1 * 72 items