Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Japan
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
ADF Volumetric Solution
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
Catalog # 305319.2500
Supplier:
VWR Chemicals
CAS Number:
Certificates
Expand image
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
Catalog # 305319.2500
Supplier:
VWR Chemicals
CAS Number:
Certificates
Specifications
パックタイプ:
Plastic bottle
Pk:
2,5 L
Seq:
5096
Specifications
About this item
Product Details
Documentation
:
Product Details
Documentation