J.T.Baker® BAKERBOND spe™ Florisil, SPE Columns
Catalog # 7213-27
Supplier: Avantor
Specifications
- 粒子サイズ:73 - 140 µm
- 容量:6 ml
- 説明:Jumbo pack column
- Seq:05
- Bed weight:1000 mg
- Phase:Florisil
- Pk:250
Specifications
About this item
BAKERBOND spe™ columns are disposable PP or glass columns pre-packed with high capacity sorbents contained between two 20 µm PE frits.
- Silica based
- Extraction of compounds from sample solution volumes ranging from a few hundred µl up to 1 L
- Processing by vacuum, positive pressure or centrifugation
General applications: Low to moderately polar analytes from non aqueous solutions.
These SPE columns can be used in adsorption mode.
Packaging: Certain columns are available in a foil-lined fibre drum called the Jumbo Pack. The columns are loose packed in the foil liner to eliminate the inconvenience of opening individual pouches.