Orders LinkContactUsLinkComponent
VWR® Basic, Aprons, PE
VWR® Basic, Aprons, PE
Supplier:   VWR Collection
VWR® Basic, Aprons, PE
Supplier:  VWR Collection
Certificates
VWR® Basic, Aprons, PE
VWR® Basic, Aprons, PE
2 Options Available Below
Product Details & Documents
Disposable aprons made of PE film.
  • Free from natural rubber latex
  • Available loose or blocked
L×W: 1250×800 mm / 1400×800 mm / 1600×800 mm
Thickness: 20, 50 or 100 µm
Product Family Options
50 pieces per bag, 10 bags per carton, thickness: 20 µm
Product Information
  • 説明
    Colour
    W×L
    Availability
    Price
  • Blocked
    Blue
    800×1250 mm
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
50 pieces per bag, 5 bags per carton, thickness: 50 µm
Product Information
  • 説明
    Colour
    W×L
    Availability
    Price
  • Blocked
    Blue
    800×1250 mm
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items