VWR®, ビジターコート、PP、防水
Supplier: VWR Collection
About this item
These disposable, waterproof visitor coats, with collar, are made from non woven, laminated PP fabric.
- Press stud closure
- Elastic wrists
- Two pockets (one breast pocket, one side pocket)
- Free from natural rubber latex
Weight: 48 g/m²
Packaging: 1 piece per bag, 50 bags per case.
3 Options Available Below
Product Details & Documents
These disposable, waterproof visitor coats, with collar, are made from non woven, laminated PP fabric.
- Press stud closure
- Elastic wrists
- Two pockets (one breast pocket, one side pocket)
- Free from natural rubber latex
Weight: 48 g/m²
Packaging: 1 piece per bag, 50 bags per case.
Documents
Food contact certificate F
Additional Product Details
VWR® Non Regulated Disposables for Production
Product Literature
Specifications for Product Family
Specifications
- Catalog No:
- 113-8983
- 113-8984
- 113-8994
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Pockets:
- With pockets
- With pockets
- With pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Garment application:
- General Purpose|Laboratory
- General Purpose|Laboratory
- General Purpose|Laboratory
- Weight:
- 48 g/m²
- 48 g/m²
- 48 g/m²
Specifications
Product Family Options
Product Information
- Colour衣服のサイズAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.113-8983Closure typePress studs / snapsPocketsWith pocketsGenderUnisexGarment applicationGeneral Purpose|LaboratoryWeight48 g/m² - Specifications:
More
Specifications
Cat. No.113-8984Closure typePress studs / snapsPocketsWith pocketsGenderUnisexGarment applicationGeneral Purpose|LaboratoryWeight48 g/m² - Specifications:
More
Specifications
Cat. No.113-8994Closure typePress studs / snapsPocketsWith pocketsGenderUnisexGarment applicationGeneral Purpose|LaboratoryWeight48 g/m²