Orders LinkContactUsLinkComponent
VWR® Basic, ビジターコート、SPP
VWR® Basic, ビジターコート、SPP
VWR® Basic, ビジターコート、SPP
Supplier:  VWR Collection
Certificates
VWR® Basic, ビジターコート、SPP
15 Options Available Below
Product Details & Documents
These disposable visitor coats, with collar, are made from non woven, SPP fabric.
  • Press stud closure
  • Elastic wrists
  • Free from natural rubber latex
Weight: 30 g/m²
Packaging: 1 piece per bag, 50 bags per case.
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-8933
  • 113-8934
  • 113-8937
  • 113-8938
  • 113-8939
  • 113-8941
  • 113-8942
  • 113-8943
  • 113-8944
  • 113-8945
  • 113-8946
  • 113-8947
  • 113-8950
  • 113-8952
  • 113-8953
  • Closure type:
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Pockets:
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Garment application:
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • Weight:
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
Specifications
Product Family Options
Product Information
  • Colour
    衣服のサイズ
    長さ
    Availability
    Price
  • White
    XS
    92 cm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8933
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²
  • White
    S
    96 cm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8934
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²
  • White
    M
    100 cm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8937
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²
  • White
    L
    104 cm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8938
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²
  • White
    XL
    108 cm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8939
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²
  • White
    XXL
    112 cm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8941
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²
  • White
    3XL
    116 cm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8942
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²
  • Blue
    XS
    92 cm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8943
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²
  • Blue
    S
    96 cm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8944
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²
  • Blue
    M
    100 cm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8945
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²
  • Blue
    L
    104 cm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8946
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²
  • Blue
    XL
    108 cm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8947
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²
  • Blue
    XXL
    112 cm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8950
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²
  • Blue
    3XL
    116 cm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8952
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²
  • Green
    XL
    108 cm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8953
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²