VWR® Basic, Overalls, Dual-Zip Closure
Supplier: VWR Collection
About this item
These disposable overalls, with collar, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Collar
- Zip closure (dual-direction)
- Elasticated wrists
- Free from natural rubber latex
Weight: 40 g/m²
Packaging: 1 piece per bag, 50 bags per case.
4 Options Available Below
Product Details & Documents
These disposable overalls, with collar, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Collar
- Zip closure (dual-direction)
- Elasticated wrists
- Free from natural rubber latex
Weight: 40 g/m²
Packaging: 1 piece per bag, 50 bags per case.
Documents
Food contact certificate F
Additional Product Details
VWR® Non Regulated Disposables for Production
Product Literature
Specifications for Product Family
Specifications
- Catalog No:
- 113-9010
- 113-9011
- 113-9012
- 113-9013
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Garment application:
- General Purpose|Laboratory|Industrial
- General Purpose|Laboratory|Industrial
- General Purpose|Laboratory|Industrial
- General Purpose|Laboratory|Industrial
- Weight:
- 40 g/m²
- 40 g/m²
- 40 g/m²
- 40 g/m²
Specifications
Product Family Options
Product Information
- Colour衣服のサイズAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.113-9010Closure typeZip closureGarment applicationGeneral Purpose|Laboratory|IndustrialWeight40 g/m² - Specifications:
More
Specifications
Cat. No.113-9011Closure typeZip closureGarment applicationGeneral Purpose|Laboratory|IndustrialWeight40 g/m² - Specifications:
More
Specifications
Cat. No.113-9012Closure typeZip closureGarment applicationGeneral Purpose|Laboratory|IndustrialWeight40 g/m² - Specifications:
More
Specifications
Cat. No.113-9013Closure typeZip closureGarment applicationGeneral Purpose|Laboratory|IndustrialWeight40 g/m²