Orders LinkContactUsLinkComponent
VWR® Basic, 作業着、ジップクロージャー、ベーシック
VWR® Basic, 作業着、ジップクロージャー、ベーシック
Supplier:   VWR Collection
VWR® Basic, 作業着、ジップクロージャー、ベーシック
Supplier:  VWR Collection
Certificates
VWR® Basic, 作業着、ジップクロージャー、ベーシック
VWR® Basic, 作業着、ジップクロージャー、ベーシック
2 Options Available Below
Product Details & Documents
These disposable frocks are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime and certain dry particulates in non hazardous environments.
  • Zip closure
  • Elastic wrists
  • Raglan sleeves
  • Free from natural rubber latex
Weight: 40 g/m²
包装: 1 piece per bag, 50 bags per case.
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-9000
  • 113-9001
  • Closure type:
  • Zip closure
  • Zip closure
  • Pockets:
  • Without pockets
  • Without pockets
  • Gender:
  • Unisex
  • Unisex
  • Garment application:
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • Weight:
  • 40 g/m²
  • 40 g/m²
Specifications
Product Family Options
Product Information
  • Colour
    衣服のサイズ
    長さ
    Availability
    Price
  • White
    XL
    108 cm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9000
    Closure type
    Zip closure
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • White
    XXL
    112 cm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-9001
    Closure type
    Zip closure
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²