Orders LinkContactUsLinkComponent
VWR® Basic Visitor Coats, Velcro® Closure
VWR® Basic Visitor Coats, Velcro® Closure
VWR® Basic Visitor Coats, Velcro® Closure
VWR® Basic Visitor Coats, Velcro® Closure
VWR® Basic Visitor Coats, Velcro® Closure
These disposable coats, with collar, are made from non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • VELCRO® closure
  • Elastic wrists
  • Free from natural rubber latex
  • Available in white and blue
Weight: 30 g/m²
Packaging: 1 piece per bag, 50 bags per case.
  • Catalog No:
  • 113-8955
  • 113-8956
  • 113-8957
  • 113-8958
  • 113-8959
  • 113-8960
  • 113-8962
  • 113-8963
  • 113-8964
  • 113-8965
  • 113-8966
  • 113-8967
  • 113-8969
  • 113-8971
  • Closure type:
  • Hook-and-loop / VELCRO®
  • Hook-and-loop / VELCRO®
  • Hook-and-loop / VELCRO®
  • Hook-and-loop / VELCRO®
  • Hook-and-loop / VELCRO®
  • Hook-and-loop / VELCRO®
  • Hook-and-loop / VELCRO®
  • Hook-and-loop / VELCRO®
  • Hook-and-loop / VELCRO®
  • Hook-and-loop / VELCRO®
  • Hook-and-loop / VELCRO®
  • Hook-and-loop / VELCRO®
  • Hook-and-loop / VELCRO®
  • Hook-and-loop / VELCRO®
  • Pockets:
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Garment application:
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Weight:
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
Product Family Options
  • Colour
    衣服のサイズ
    長さ
  • White
    XS
    92 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8955
    Closure type
    Hook-and-loop / VELCRO®
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Laboratory
    Weight
    30 g/m²
  • White
    S
    96 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8956
    Closure type
    Hook-and-loop / VELCRO®
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Laboratory
    Weight
    30 g/m²
  • White
    M
    100 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8957
    Closure type
    Hook-and-loop / VELCRO®
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Laboratory
    Weight
    30 g/m²
  • White
    L
    104 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8958
    Closure type
    Hook-and-loop / VELCRO®
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Laboratory
    Weight
    30 g/m²
  • White
    XL
    108 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8959
    Closure type
    Hook-and-loop / VELCRO®
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Laboratory
    Weight
    30 g/m²
  • White
    XXL
    112 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8960
    Closure type
    Hook-and-loop / VELCRO®
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Laboratory
    Weight
    30 g/m²
  • White
    3XL
    116 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8962
    Closure type
    Hook-and-loop / VELCRO®
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Laboratory
    Weight
    30 g/m²
  • Blue
    XS
    92 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8963
    Closure type
    Hook-and-loop / VELCRO®
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Laboratory
    Weight
    30 g/m²
  • Blue
    S
    96 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8964
    Closure type
    Hook-and-loop / VELCRO®
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Laboratory
    Weight
    30 g/m²
  • Blue
    M
    100 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8965
    Closure type
    Hook-and-loop / VELCRO®
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Laboratory
    Weight
    30 g/m²
  • Blue
    L
    104 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8966
    Closure type
    Hook-and-loop / VELCRO®
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Laboratory
    Weight
    30 g/m²
  • Blue
    XL
    108 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8967
    Closure type
    Hook-and-loop / VELCRO®
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Laboratory
    Weight
    30 g/m²
  • Blue
    XXL
    112 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8969
    Closure type
    Hook-and-loop / VELCRO®
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Laboratory
    Weight
    30 g/m²
  • Blue
    3XL
    116 cm
    1 * 50 items
    1 * 50 items
    Specifications
    Cat. No.
    113-8971
    Closure type
    Hook-and-loop / VELCRO®
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Laboratory
    Weight
    30 g/m²