Orders LinkContactUsLinkComponent
VWR® Basic, シューズカバー、SPP
VWR® Basic, シューズカバー、SPP
Supplier:   VWR Collection
VWR® Basic, シューズカバー、SPP
Supplier:  VWR Collection
Certificates
VWR® Basic, シューズカバー、SPP
VWR® Basic, シューズカバー、SPP
3 Options Available Below
Product Details & Documents
These disposable shoe covers are made from a non woven SPP fabric.
  • Free from natural rubber latex
  • Available with anti-skid sole
L×H: 410×170 mm
Weight: 40 g/m²
* One size fits all
包装: 10 pieces per bundle, 10 bundles per bag, 3 bags per case.
Product Family Options
Product Information
  • 説明
    Colour
    靴のサイズ
    高さ
    長さ
    Availability
    Price
  • Standard
    White
    U*
    170 mm
    410 mm
    Log in for availability
    Log in for availability
    1 * 300 items
    1 * 300 items
  • Standard
    Blue
    U*
    170 mm
    410 mm
    Log in for availability
    Log in for availability
    1 * 300 items
    1 * 300 items
  • Anti-skid
    Blue
    U*
    170 mm
    410 mm
    Log in for availability
    Log in for availability
    1 * 300 items
    1 * 300 items