Orders LinkContactUsLinkComponent
VWR®, バイザー付きブーファントキャップ、
VWR®, バイザー付きブーファントキャップ、
VWR®, バイザー付きブーファントキャップ、
VWR®, バイザー付きブーファントキャップ、
These disposable caps, with peak, are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Free from natural rubber latex
  • Available in white and blue
Weight: 10 g/m² (cap), 100 g/m² (peak)
 : 100 pieces per bag, 10 bags per case.
Product Family Options
  • Colour
  • White
    1 * 1.000 items
    1 * 1.000 items
  • Blue