Orders LinkContactUsLinkComponent
VWR® Basic, ヒゲカバー、
VWR® Basic, ヒゲカバー、
VWR® Basic, ヒゲカバー、
Supplier:  VWR Collection
Certificates
VWR® Basic, ヒゲカバー、
2 Options Available Below
Product Details & Documents
This disposable beard cover is made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Free from natural rubber latex
  • Available in white and blue
Weight: 10 g/m²
Packaging: 100 pieces/bag, 10 bags/case.
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-8902
  • 113-8903
  • Weight:
  • 10 g/m²
Specifications
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • White
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8902
    Weight
    10 g/m²
  • Blue
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8903
    Weight