Orders LinkContactUsLinkComponent
VWR® Basic, モブキャップ、検出可能、ベーシック
VWR® Basic, モブキャップ、検出可能、ベーシック
Supplier:   VWR Collection
VWR® Basic, モブキャップ、検出可能、ベーシック
Supplier:  VWR Collection
Certificates
VWR® Basic, モブキャップ、検出可能、ベーシック
VWR® Basic, モブキャップ、検出可能、ベーシック
4 Options Available Below
Product Details & Documents
These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Free from natural rubber latex
  • Double elastic
  • Metal detectable, therefore, ideal for use in the food processing industry
Weight: 10 g/m²
Packaging: 100 pieces per bag, 10 bags per case.
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-8895
  • 113-8896
  • 113-8897
  • 113-8898
  • Garment application:
  • General Purpose|Food
  • General Purpose|Food
  • General Purpose|Food
  • General Purpose|Food
  • Weight:
  • 10 g/m²
  • 10 g/m²
  • 10 g/m²
  • 10 g/m²
Specifications
Product Family Options
Product Information
  • Colour
    直径
    Availability
    Price
  • White
    530 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8895
    Garment application
    General Purpose|Food
    Weight
    10 g/m²
  • Blue
    530 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8896
    Garment application
    General Purpose|Food
    Weight
    10 g/m²
  • Green
    530 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8897
    Garment application
    General Purpose|Food
    Weight
    10 g/m²
  • Red
    530 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8898
    Garment application
    General Purpose|Food
    Weight
    10 g/m²