Orders LinkContactUsLinkComponent
VWR® Basic, モブキャップ、シングルエラスティックベーシック
Supplier:   VWR Collection
VWR® Basic, モブキャップ、シングルエラスティックベーシック
Supplier:  VWR Collection
Certificates
VWR® Basic, モブキャップ、シングルエラスティックベーシック
VWR® Basic, モブキャップ、シングルエラスティックベーシック
12 Options Available Below
Product Details & Documents
These disposable pleated caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Free from natural rubber latex
  • Single elastic
  • Available in a wide range of colours
Weight: 10 g/m²
 : 100 pieces per bag, 10 bags per case.
Product Family Options
Product Information
  • Colour
    直径
    Availability
    Price
  • White
    530 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • Blue
    530 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • Green
    530 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • White
    580 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • Blue
    580 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • Green
    580 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • Red
    580 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • Yellow
    580 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • Orange
    580 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • Red
    530 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • Yellow
    530 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • Orange
    530 mm