Orders LinkContactUsLinkComponent
VWR®, ブロックヒーター、デジタル
VWR®, ブロックヒーター、デジタル
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, ブロックヒーター、デジタル
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, ブロックヒーター、デジタル
4 Options Available Below
Product Details & Documents
Multi-purpose, high wattage, constant temperature, VWR® digital block heaters are economical, versatile and compact. These digital models provide reproducable results, exceptional temperature uniformity, and are ideal for a wide range of applications. Temperature calibration mode allows the user to calibrate the unit to an external temperature device. In timed mode, an alarm will sound when the time reaches zero and the unit reaches set point temperature.
  • Compact design with small footprint
  • Temperature range from ambient +5 to 120 °C
  • 'Hot' symbol warning light is illuminated when the temperature is above 40 °C
  • Accommodates VWR® interchangeable modular heating blocks for tubes from 0,2 to 50 ml capacity
  • Can be used at 18 to 33 °C, with 20 to 80% relative humidity, non condensing
These multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, denaturing proteins, DNA applications, ELISA, melting/boiling points, and a variety of other laboratory procedures.
Microprocessor control: PID microprocessor temperature control, with optional external RTD probe, offers a temperature stability as low as ±0,1 °C, with a temperature uniformity as low as ±0,1 °C. Samples are heated to temperature quickly and accurately, heat-up time to 100 ºC is 16 minutes. Temperature is adjusted in 0,1 °C increments.
LED display: Touch pad controls with easy to read, independent LED displays for temperature and time. Shows set point and actual temperatures. Timer displays elapsed time or, when programmed to user-defined limit, will shut off unit when time reaches zero.
Units have separate interchangeable modular heating blocks, which accommodate various sample containers such as microtubes, centrifuge tubes, vials, microplates and PCR strips or tubes. Blocks include a thermometer well, and feature close 'tube and block' contact for maximum heat retention. All dry block heaters include a built-in support rod holder for mounting the probe kit.
Certifications: TUV listed, RoHS compliant.
注文情報: Supplied without heating blocks, please order these separately. Optional external temperature-probe kit monitors actual block or sample temperature.
注意: To avoid a possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed for use as a dry bath/incubator only.
* RT = Ambient
Specifications for Product Family
Specifications
  • Catalog No:
  • 460-0349
  • 460-0350
  • 460-0351
  • 460-0352
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
Specifications
Product Family Options
Product Information
  • Type
    No. of blocks
    Heating capacity
    温度範囲
    Temp. stability
    W×D×H
    Weight
    Availability
    Price
  • Block heater, digital
    1
    0,10 kW
    RT* +5...120 °C
    ±0,2 °C
    196×269×117 mm
    2,3 kg
    1 * 1 items
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    460-0349
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • Block heater, digital
    2
    0,19 kW
    RT* +5...120 °C
    ±0,2 °C
    196×348×117 mm
    2,6 kg
    1 * 1 items
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    460-0350
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • Block heater, digital
    4
    0,27 kW
    RT* +5...120 °C
    ±0,2 °C
    201×424×104 mm
    4 kg
    1 * 1 items
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    460-0351
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • Block heater, digital
    6
    0,42 kW
    RT* +5...120 °C
    ±0,2 °C
    201×526×104 mm
    4,5 kg
    1 * 1 items
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    460-0352
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
Accessories
Product Information
  • Catalog #
    説明
    Well size
    穴の数
    Availability
    Price
  • 460-3237
    Single block, 2×6 cuvettes (12,5 mm)
    - mm
    -
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3243
    Single block
    Ø 19,1 mm
    17/18 mm round bottom tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3224
    Single block, 10×8-PCR tube strips
    Ø 6,4 mm
    0,2 ml tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3285
    Single block
    Ø 25,8 mm
    25 mm vials
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3280
    Single block
    Ø 15,8 mm
    15 mm vials
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3287
    Single block
    Ø 16,4 mm
    16 mm vials
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3213
    Single block
    Ø 8,3 mm
    6 mm round bottom tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3282
    Single block
    Ø 19,7 mm
    19 mm vials
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3279
    Single block
    Ø 12,7 mm
    12 mm vials
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3222
    Single block
    Ø 35,0 mm
    35 mm round bottom tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3218
    Single block
    Ø 21,0 mm
    20 mm round bottom tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    穴の数
    Availability
    Price
  • 460-3220
    Single block, test tube combination 6 mm, 12/13 mm, 25 mm
    Ø 8,3 / Ø 13,9 / Ø 26,2 mm
    6/5/3
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    穴の数
    Availability
    Price
  • 460-3248
    Single block, microtube combination 0,5 ml, 1,5 ml, 2,0 ml
    Ø 7,9 / Ø 11,1 / Ø 11,5 mm
    6/10/5
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3215
    Single block
    Ø 13,9 mm
    12/13 mm round bottom tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3214
    Single block
    Ø 10,7 mm
    10 mm round bottom tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3212
    Single block
    Ø 11,5 mm
    1,5 ml Eppendorf microtubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3216
    Single block
    Ø 13,9 mm
    12/13 mm round bottom tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3219
    Single block
    Ø 26,2 mm
    25 mm round bottom tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3284
    Single block
    Ø 23,8 mm
    23 mm vials
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • VWRU14216-212
    Single block
    Ø 19,1 mm
    17/18 mm round bottom tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3221
    Single block
    Ø 17,1 mm
    15 ml conical tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    W×D×H
    Availability
    Price
  • 460-3236
    Single block
    - mm
    95×76×51 mm
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    W×D×H
    Availability
    Price
  • 460-3239
    Double block
    - mm
    95×152×57 mm
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    穴の数
    Availability
    Price
  • 460-3247
    Single block, centrifuge tube combination 1,5 ml, 15 ml, 50 ml
    Ø 11,1 / Ø 17,1 / Ø 29,0 mm
    4/3/2
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3225
    Single block, 64× individual PCR tubes
    Ø 6,4 mm
    0,2 ml tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3235
    Single block
    Ø 11,1 mm
    1,5 ml VWR microtubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3286
    Single block
    Ø 28,8 mm
    28 mm vials
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3217
    Single block
    Ø 17,5 mm
    15/16 mm round bottom tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3211
    Single block
    Ø 7,9 mm
    0,5 ml PCR tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3223
    Single block
    Ø 29,0 mm
    50 ml conical tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3234
    Single block
    Ø 6,3 mm
    Round bottomed tubes, 6 mm Ø
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3283
    Single block
    Ø 21,7 mm
    21 mm vials
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3226
    Double block, 1×96-well PCR plate
    Ø 6,4 mm
    0,2 ml tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3281
    Single block
    Ø 17,8 mm
    17 mm vials
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3238
    Double block
    - mm
    96- or 384-well plate
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3245
    Single block
    Ø 11,5 mm
    2,0 ml VWR/Eppendorf microtubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Well size
    For
    Availability
    Price
  • 460-3246
    Single block
    Ø 10,9 mm
    2,0 ml Corning microtubes