Orders LinkContactUsLinkComponent
VWR®, エレクトロポレーション用キュベット
VWR®, エレクトロポレーション用キュベット
VWR®, エレクトロポレーション用キュベット
Supplier:  VWR Collection
Certificates
VWR®, エレクトロポレーション用キュベット
VWR®, エレクトロポレーション用キュベット
3 Options Available Below
Product Details & Documents
These electroporation cuvettes are made from clear, medical grade polycarbonate and are compatible with all common commercially available electroporators.
  • Polished aluminium lowers arcing frequency
  • Colour-coded caps for easy identification of gap sizes
  • Round caps for easy, single-handed cap removal
Ordering information: Available in three gap sizes to meet common application requirements: 1 mm for highest field strength, suitable for bacteria; 2 mm gap for intermediate requirements; 4 mm gap for lowest field strength, suitable for mammalian cells and some plant cells.
Packaging: Sterilised by gamma irradiation and individually packed for assured sterility.
Product Family Options
Product Information
  • 説明
    容量
    Availability
    Price
  • Cuvettes with 1 mm gap size, grey cap
    20 - 90 µl
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
  • Cuvettes with 2 mm gap size, blue cap
    40 - 400 µl
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
  • Cuvettes with 4 mm gap size, yellow cap
    80 - 800 µl