Orders LinkContactUsLinkComponent
VWR® Star Tec, 実験用椅子
   Sustainable Options Available
Supplier:   VWR Collection
VWR® Star Tec, 実験用椅子
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR® Star Tec, 実験用椅子
6 Options Available Below
Product Details & Documents
These laboratory chairs, with seat and back in black upholstery artificial leather, are soft and comfortable. The permanent backrest follows all movements and supports dynamic sitting in laboratories. Extra large and ergonomically designed backrest with integrated lumbar support, the upholstery pads on the backrest provide maxiumum seating comfort and optimal microclimate, the tapered backrest in the upper area offers elbow room for flexible work in the laboratory and maximum freedom of movement.
  • Washable, resistant to desinfectants
  • Robust and easy to clean
  • Backrest adjustable in height and inclination
  • Seat inclination adjustment supports a forward angled working position which is typical for laboratory work
  • Every function can be operated comfortable and easily in a sitting position
  • Pratical handle on the backrest provides maximum mobility and easy handling
  • Solid plastic black star base
Warranty: 10-year long-term guarantee, free shipping of spare parts during the whole period of validity.
 : Suitable for use in biosafety laboratories BSL 1 and BSL 2, GMP conformity.
Specifications for Product Family
Specifications
  • Catalog No:
  • 132-1390
  • 132-1391
  • 132-1392
  • 132-1393
  • 132-1394
  • 132-1395
  • Environmentally Preferable:
  • Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Environmental certification:
  • ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
  • ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
  • ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
  • ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
  • ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
  • ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
Specifications
Product Family Options
Product Information
  • 説明
    高さ
    Foot style
    Foot ring
    Availability
    Price
  • Laboratory chair Star Tec, with black upholstery artificial leather and blue coloured handle
    450 - 630 mm
    Soft castors for hard floors
    1 * 1 items
    Specifications:
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    132-1390
    Environmentally Preferable
    Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    Environmental certification
    ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Laboratory chair Star Tec, with black upholstery artificial leather and grey coloured handle
    450 - 630 mm
    Soft castors for hard floors
    1 * 1 items
    Specifications:
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    132-1391
    Environmentally Preferable
    Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    Environmental certification
    ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Laboratory chair Star Tec, with black upholstery artificial leather and blue coloured handle
    590 - 860 mm
    Large glides, plastic
    Height adjustable, chrome plated
    1 * 1 items
    Specifications:
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    132-1392
    Environmentally Preferable
    Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    Environmental certification
    ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Laboratory chair Star Tec, with black upholstery artificial leather and grey coloured handle
    590 - 860 mm
    Large glides, plastic
    Height adjustable, chrome plated
    1 * 1 items
    Specifications:
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    132-1393
    Environmentally Preferable
    Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    Environmental certification
    ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Laboratory chair Star Tec, with black upholstery artificial leather and blue coloured handle
    620 - 890 mm
    Castors, Stop&Go
    Height adjustable, chrome plated
    1 * 1 items
    Specifications:
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    132-1394
    Environmentally Preferable
    Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    Environmental certification
    ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Laboratory chair Star Tec, with black upholstery artificial leather and grey coloured handle
    620 - 890 mm
    Castors, Stop&Go
    Height adjustable, chrome plated
    1 * 1 items
    Specifications:
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    132-1395
    Environmentally Preferable
    Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    Environmental certification
    ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
Accessories
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 132-0702
    4-D-multifunctional armrest: Height, depth and width adjustable as well as pivoted
    Log in for availability
    1 * 2 items