マグネット式スターラーバー、Spinwedge® (スピンウェッジ)
9,5 mm, 25 mm
Catalog # VWRU58949-232
Supplier: VWR Collection
Jump to:
Product Details & Documents
These magnetic stirring bars provide strong turbulence at fairly low speeds and are well suited for churning sediment or dissolving salts.
- Wedge shape is designed to plow up sediment for thorough dispersion
- Sharply angled broad face provides strong mixing action, perfect for viscous solutions
- Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
- Autoclave safe
Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert.
Specifications
- 直径:25 mm
- 長さ:9,5 mm
- Seq:01