Orders LinkContactUsLinkComponent
UVハンドヘルドランプ、VWR®製
UVハンドヘルドランプ、VWR®製
Supplier:   VWR Collection
UVハンドヘルドランプ、VWR®製
Supplier:  VWR Collection
Certificates
UVハンドヘルドランプ、VWR®製
UVハンドヘルドランプ、VWR®製
2 Options Available Below
Product Details & Documents
These UV lamps have many uses depending on the wavelength of emitted UV light.
  • Dual UV lamps
  • Use 6-watt tubes and are manufactured of lightweight rugged aluminum with a scratch-resistant powder paint
  • UV-A hand lamp features two 365 nm longwave UV tubes for brilliant fluorescence
Applications for 365 nm UV include water testing, quality control inspection, non destructive testing and sanitation. The 302 nm UV can be used in UV curing, gel viewing, and mineralogy.
Product Family Options
Product Information
  • 説明
    モデル
    波長
    Availability
    Price
  • UV handheld lamp, 115 V
    UV-AB
    302/365 nm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $1,863.05
    $1,863.05
    1 * 1 items
       $1,863.05
  • UV handheld lamp, 230 V
    UV-AB
    302/365 nm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $1,994.16
    $1,994.16
    1 * 1 items
       $1,994.16