Orders LinkContactUsLinkComponent
マイクロチューブラック、80箇所
マイクロチューブラック、80箇所
Supplier:   VWR Collection
マイクロチューブラック、80箇所
Supplier:  VWR Collection
Certificates
マイクロチューブラック、80箇所
マイクロチューブラック、80箇所
3 Options Available Below
Product Details & Documents
Fraction collector plate style rack made of PP.
  • Moulded-in coordinates to aid orientation
  • Conical shaped holes (Ø 11 mm) to hold microtubes securely
  • Easily fill and transfer 80 tubes in compact arrangement (5×16 array)
Product Family Options
Product Information
  • 説明
    Colour
    穴の数
    For tubes
    W×D×H
    Availability
    Price
  • VWR® 80-well microtube rack
    Assorted
    80 (5×16)
    1,5/2,0 ml
    225×67×28 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items - $42.71
    $42.71
    1 * 5 items
       $42.71
  • VWR® 80-well microtube rack
    Pink
    80 (5×16)
    1,5/2,0 ml
    225×67×28 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items - $43.43
    $43.43
    1 * 5 items
       $43.43
  • VWR® 80-well microtube rack
    Yellow
    80 (5×16)
    1,5/2,0 ml
    225×67×28 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items - $43.43
    $43.43
    1 * 5 items
       $43.43