VWR® TraceClean®, Water sampling bottles, hdpe, wide neck
Supplier: VWR Collection
Certificates
About this item
Bottles are used in collection of water samples for metals analyses and a variety of inorganic analytes including cyanide, sulphide, sulphate, chloride, and ion chromatography.
- HDPE construction
- Wide-mouthed design
Processed (PC) and Quality-Assured® (QA) packers follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A "Specifications and Guidance for Contaminant-Free Sample Containers." QA packers include a certificate of analysis in every case. QA packers also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) packers do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.
Delivery: Supplied with polyfoam-lined caps.
Frequently Bought Together