Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Japan
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
インキュベーター
Compact Benchtop Incubators
Accessories for VWR® Personal Sized Incubator
Accessories for VWR® Personal Sized Incubator
Accessories for VWR® Personal Sized Incubator
Catalog # VWRU97025-636
Supplier:
VWR Collection
Expand image
Accessories for VWR® Personal Sized Incubator
Catalog # VWRU97025-636
Supplier:
VWR Collection
Specifications
説明:
Thermometer, –20...110 °C
Seq:
02
Pk:
1
For:
Use with small VWR personal incubator
Specifications
About this item
Product Details
Product Details