スワブアプリケーター、VWR®
Swab applicators, 152 mm
Catalog # VWRU470019-172
Supplier: VWR Collection
Jump to:
Product Details & Documents
This sterile cotton-tipped applicator spreads a confluent lawn of bacteria without breaking the agar surface.
- Easy sub-culturing and bacterial transfers
包装: Individual.
注意: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- 説明:Swab applicators
- 長さ:152 mm