Orders LinkContactUsLinkComponent
VWR®, シングルウェル細胞培養プレート
VWR®, シングルウェル細胞培養プレート
VWR®, シングルウェル細胞培養プレート
VWR®, シングルウェル細胞培養プレート
Single-well cell culture plates, made from clear PS, are available plasma-treated for consistent cell attachment and growth, or non-treated for use in applications where cell attachment is not desired.
  • Increased internal plate dimension, providing larger area for cell growth
  • Standard SBS footprint makes them compatible with standard equipment and automated systems
  • Free from DNase, RNase, and human DNA, certified non-pyrogenic, non-cytotoxic
  • Sterilised by electron beam irradiation
TC-treated: These dishes are vacuum-gas plasma or corona treated for consistent cell attachment and growth.
Non-treated: Non-treated cell and tissue culture dishes are ideal for applications where cell attachment is not desired.
Temperature range: –20 to +50 °C.
 : ISO 13485:2003, ISO 9001:2008.
  • Catalog No:
  • 734-2966
  • 734-2977
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
Product Family Options
  • 説明
    ウェルの数
    Culture area
    Recommended working volume
    Well volume
  • Single-well plate, TC treated
    1
    97 cm²
    35 ml
    73 ml
    1 * 100 items
    1 * 100 items
      Low Manufacturing Impact
    Specifications
    Cat. No.
    734-2966
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • Single-well plate, non treated
    1
    97 cm²
    35 ml
    73 ml
    1 * 100 items
    1 * 100 items
      Low Manufacturing Impact
    Specifications
    Cat. No.
    734-2977
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M