VWR®, セルリフター
Flat blade cell lifter for manual harvesting of cells, manufactured from high grade PE. Designed for removing cells from flasks or dishes.
- Bevelled edge blade on one end and narrow blade or J-hook on the other
- Sterilised by electron beam irradiation
- Free from DNase, RNase and non-pyrogenic
Flat blade cell lifter for manual harvesting of cells, manufactured from high grade PE. Designed for removing cells from flasks or dishes.
- Bevelled edge blade on one end and narrow blade or J-hook on the other
- Sterilised by electron beam irradiation
- Free from DNase, RNase and non-pyrogenic
- Catalog No:
- 734-2978
- 734-2979
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Product Family Options
- 説明総長
Low Manufacturing Impact Specifications
Cat. No.734-2978Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact Specifications
Cat. No.734-2979Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact