VWR®, デジタルタイマー、1チャンネル、
White, 0,01 %
Catalog # 609-0224
Supplier: VWR Collection
Jump to:
Product Details & Documents
This single-channel timer can be programmed in countdown or count up mode.
- Timing capacity: 99 minutes, 59 seconds
- Resolution is 1 second
- Alarm and memory function
- Big digit display
納品情報: Supplied with magnet, clip and stand.
Specifications
- Colour:White
- Accuracy:0,01 %