ブレンダーバッグ、FILTRA-BAG®(フィルトラバッグ
Supplier: VWR Collection
Certificates
About this item
These FILTRA-BAG® blender bags are designed to simplify taking an aliquot when working with samples which contain large amounts of residue and/or semi-solid/solid substances.
- Made with a PE/PET outer shell which practically eliminates the possibility of piercing the bag wall during blending
- Prevents pipette blockage as well as uneven plate distribution
- Dividing filter membrane has 1840 holes per square inch with a pore size of approximately 330 microns
Frequently Bought Together