マイクロスパチュラ、 両端
Supplier: VWR Collection
Jump to:
About this item
These micro spatulas are double-ended and made of stainless steel.
2 Options Available Below
Product Details & Documents
These micro spatulas are double-ended and made of stainless steel.
Product Family Options
Product Information
- 説明Blade lengthBlade widthAvailabilityPrice
- Micro spatula with curved, square-cut sharpened tip and rounded spoon at other end165 mm3 mmLog in for availability1 * 1 items$17.64
- Micro spatula with bent, sharpened tip and V-shaped sharpened spoon at other end170 mm2 mmLog in for availability1 * 1 items$18.08