VWR®, カラーフロスト顕微鏡スライド
About this item
Microscope slides recommended for haematoxylin and eosin (H&E) stain and hybridisation on frozen sections in histopathology examination.
- Smooth edges for operational safety
- Sticky surface for stronger adhesion than with poly-l-lysine
- Marking with conventional labelling system, pencils, marker pens and thermal transfer printers
Recommended for hematoxylin and eosin (H&E) stain, immunohistochemistry (IHC) and hybridisation on frozen sections in histopathology examination.
Packaging: 50 pieces per box. 50 boxes per case.
Caution: To be stored in dry and proper laboratory temperature conditions − shelf life of 60 months.
5 Options Available Below
Product Details & Documents
Microscope slides recommended for haematoxylin and eosin (H&E) stain and hybridisation on frozen sections in histopathology examination.
- Smooth edges for operational safety
- Sticky surface for stronger adhesion than with poly-l-lysine
- Marking with conventional labelling system, pencils, marker pens and thermal transfer printers
Recommended for hematoxylin and eosin (H&E) stain, immunohistochemistry (IHC) and hybridisation on frozen sections in histopathology examination.
Packaging: 50 pieces per box. 50 boxes per case.
Caution: To be stored in dry and proper laboratory temperature conditions − shelf life of 60 months.
Product Family Options
Product Information
- 説明Colour厚さ幅×奥行きAvailabilityPrice
- Colour-frosted microscope slidesBlue1,0 (-0,05/+0,1) mm76×25 mmLog in for availability1 * 2.500 itemsSpecifications:
More
Specifications
Cat. No.631-1169 - Colour-frosted microscope slidesGreen1,0 (-0,05/+0,1) mm76×25 mmLog in for availability1 * 2.500 itemsSpecifications:
More
Specifications
Cat. No.631-1172 - Colour-frosted microscope slidesOrange1,0 (-0,05/+0,1) mm76×25 mmLog in for availability1 * 2.500 itemsSpecifications:
More
Specifications
Cat. No.631-1173 - Colour-frosted microscope slidesPink1,0 (-0,05/+0,1) mm76×25 mmLog in for availability1 * 2.500 itemsSpecifications:
More
Specifications
Cat. No.631-1175 - Colour-frosted microscope slidesYellow1,0 (-0,05/+0,1) mm76×25 mmLog in for availability1 * 2.500 itemsSpecifications:
More
Specifications
Cat. No.631-1176